Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151717.2 | XP_015007203.1 | 497 | 74 | ||
XM_015151718.2 | XP_015007204.1 | 441 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |