Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028848176.1 | XP_028704009.1 | 1075 | 137 | ||
XM_028848177.1 | XP_028704010.1 | 1066 | 137 | |||
XM_028848178.1 | XP_028704011.1 | 872 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |