Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: SLC9C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015129854.2 | XP_014985340.2 | 3302 | 161 | ||
XM_015129855.2 | XP_014985341.2 | 3302 | 161 | |||
XM_015129857.2 | XP_014985343.2 | 3203 | 161 | |||
XM_015129859.2 | XP_014985345.2 | 3140 | 161 | |||
XM_015129862.2 | XP_014985348.1 | 2692 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |