Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC106996466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842643.1 | XP_028698476.1 | 539 | 121 | ||
XM_028842644.1 | XP_028698477.1 | 539 | 121 | |||
GenBank mRNA | JV492735.1 | - | 228 | 121 | ||
JV604152.1 | - | 138 | 121 |
Gene Symbol: LOC707689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028850021.1 | XP_028705854.1 | 217 | 121 | ||
GenBank mRNA | JV671218.1 | - | 136 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |