Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266655.1 | NP_001253584.1 | 425 | 171 | ||
XM_015111694.2 | XP_014967180.1 | 460 | 171 | |||
GenBank mRNA | JU474575.1 | - | 314 | 171 | ||
JV046365.1 | - | 461 | 171 | |||
JV635866.1 | - | 425 | 171 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |