Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRACDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832203.1 | XP_028688036.1 | 972 | 100 | ||
XM_028832204.1 | XP_028688037.1 | 538 | 100 | |||
XM_028832205.1 | XP_028688038.1 | 564 | 100 | |||
XM_028832206.1 | XP_028688039.1 | 374 | 100 | |||
XM_028832207.1 | XP_028688040.1 | 341 | 100 | |||
XM_028832208.1 | XP_028688041.1 | 541 | 100 | |||
XM_028832209.1 | XP_028688042.1 | 618 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |