Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRACDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832203.1 | XP_028688036.1 | 1109 | 77 | ||
XM_028832204.1 | XP_028688037.1 | 675 | 77 | |||
XM_028832205.1 | XP_028688038.1 | 701 | 77 | |||
XM_028832206.1 | XP_028688039.1 | 511 | 77 | |||
XM_028832207.1 | XP_028688040.1 | 478 | 77 | |||
XM_028832208.1 | XP_028688041.1 | 678 | 77 | |||
XM_028832209.1 | XP_028688042.1 | 755 | 77 |