Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206975.1 | NP_001193904.1 | 464 | 132 | ||
GenBank mRNA | JU320670.1 | - | 177 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Macaca mulatta Inventoried |