Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR1M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194210.3 | NP_001181139.3 | 554 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |