Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM78B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028849989.1 | XP_028705822.1 | 770 | 97 | ||
GenBank mRNA | JV606867.1 | - | 681 | 97 | ||
JV674020.1 | - | 160 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |