Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135426.2 | XP_014990912.2 | 1615 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |