Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114260.3 | XP_001114260.2 | 1512 | 106 | ||
XM_001114298.3 | XP_001114298.2 | 1390 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |