Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042647.1 | NP_001036112.1 | 1042 | 126 | ||
GenBank mRNA | AF084226.1 | - | 1042 | 126 | ||
AF084227.1 | - | 1042 | 126 | |||
DQ887996.1 | - | 1042 | 126 | |||
DQ887998.1 | - | 1042 | 126 | |||
DQ887999.1 | - | 1042 | 126 | |||
DQ888003.1 | - | 1042 | 126 | |||
DQ888004.1 | - | 1042 | 126 | |||
DQ888017.1 | - | 1042 | 126 | |||
DQ888024.1 | - | 1042 | 126 | |||
DQ888026.1 | - | 1042 | 126 | |||
DQ888027.1 | - | 1042 | 126 | |||
DQ888031.1 | - | 1042 | 126 | |||
DQ888035.1 | - | 1042 | 126 | |||
DQ888036.1 | - | 1042 | 126 | |||
DQ888037.1 | - | 1042 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |