Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265629.1 | NP_001252558.1 | 153 | 117 | ||
GenBank mRNA | JV565030.1 | - | 153 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta Inventoried |