Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015132526.2 | XP_014988012.1 | 597 | 100 | ||
GenBank mRNA | JV600231.1 | - | 130 | 100 | ||
JV649978.1 | - | 78 | 100 | |||
JV713554.1 | - | 209 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta Inventoried |