Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB21L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194220.3 | NP_001181149.1 | 619 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |