Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198698.1 | NP_001185627.1 | 179 | 124 | ||
XM_028834209.1 | XP_028690042.1 | 381 | 124 | |||
XM_028834210.1 | XP_028690043.1 | 243 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |