Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028845939.1 | XP_028701772.1 | 3498 | 48 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |