Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261786.1 | NP_001248715.1 | 243 | 142 | ||
GenBank mRNA | JU321745.1 | - | 60 | 142 | ||
JV048184.1 | - | 243 | 142 | |||
JV636886.1 | - | 346 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |