Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TICAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130428.1 | NP_001123900.1 | 498 | 160 | ||
XM_015122534.2 | XP_014978020.2 | 779 | 160 | |||
GenBank mRNA | AB446490.1 | - | 498 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |