Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266147.1 | NP_001253076.1 | 5465 | 144 | ||
GenBank mRNA | JU473120.1 | - | 5465 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |