Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195661.1 | NP_001182590.1 | 1019 | 143 | ||
XM_015132415.2 | XP_014987901.1 | 2902 | 143 | |||
XM_028843269.1 | XP_028699102.1 | 2587 | 143 | |||
GenBank mRNA | CB229930.1 | - | 520 | 143 | ||
JU327120.1 | - | 975 | 143 | |||
JV535894.1 | - | 982 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |