Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260847.1 | NP_001247776.1 | 341 | 144 | ||
GenBank mRNA | JU336864.1 | - | 338 | 144 | ||
JU477604.1 | - | 1761 | 144 | |||
JV048545.1 | - | 341 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta Inventoried |