Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC711820 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112259.4 | XP_001112259.2 | 2291 | 117 | ||
XM_015119799.2 | XP_014975285.2 | 909 | 117 | |||
XM_028836528.1 | XP_028692361.1 | 1609 | 117 | |||
XM_028836529.1 | XP_028692362.1 | 1630 | 117 | |||
GenBank mRNA | JV664292.1 | - | 205 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |