Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194099.2 | NP_001181028.2 | 107 | 115 | ||
XM_015116395.2 | XP_014971881.2 | 128 | 115 | |||
XM_028834366.1 | XP_028690199.1 | 128 | 115 | |||
XM_028834367.1 | XP_028690200.1 | 128 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |