Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097698.4 | XP_001097698.1 | 176 | 142 | ||
GenBank mRNA | JV659485.1 | - | 64 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |