Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260920.1 | NP_001247849.1 | 257 | 152 | ||
GenBank mRNA | JU322855.1 | - | 269 | 152 | ||
JU472232.1 | - | 265 | 152 | |||
JV044834.1 | - | 257 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |