Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257443.2 | NP_001244372.1 | 297 | 156 | ||
GenBank mRNA | JU322385.1 | - | 298 | 156 | ||
JU472753.1 | - | 308 | 156 | |||
JV047363.1 | - | 311 | 156 | |||
JV547758.1 | - | 324 | 156 | |||
JV667238.1 | - | 277 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |