Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMSB4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260557.2 | NP_001247486.1 | 368 | 135 | ||
XM_028841815.1 | XP_028697648.1 | 810 | 135 | |||
GenBank mRNA | CB309232.1 | - | 371 | 135 | ||
JV048100.1 | - | 298 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |