Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148758.2 | XP_015004244.1 | 1361 | 132 | ||
XM_028827088.1 | XP_028682921.1 | 1178 | 132 | |||
XM_028827089.1 | XP_028682922.1 | 1352 | 132 | |||
XM_028827091.1 | XP_028682924.1 | 1381 | 132 | |||
GenBank mRNA | JU328418.1 | - | 1067 | 132 | ||
JU473144.1 | - | 1152 | 132 | |||
JV045237.1 | - | 1149 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |