Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032862.1 | NP_001028034.1 | 183 | 89 | ||
XM_015144691.2 | XP_015000177.1 | 316 | 89 | |||
GenBank mRNA | AF184159.1 | - | 183 | 89 | ||
AF184160.1 | - | 183 | 89 | |||
AF188268.1 | - | 222 | 89 | |||
AF188269.1 | - | 222 | 89 | |||
AF188270.1 | - | 222 | 89 | |||
JU492670.1 | - | 114 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |