Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040212.1 | NP_001035302.1 | 819 | 63 | ||
GenBank mRNA | AY635461.1 | - | 819 | 63 | ||
JV643564.1 | - | 41 | 63 |
Gene Symbol: LOC114675607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842827.1 | XP_028698660.1 | 868 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |