Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040240.1 | NP_001035330.1 | 1030 | 72 | ||
GenBank mRNA | AY952296.1 | - | 1030 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |