Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS2R46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080757.3 | NP_001074226.2 | 755 | 79 | ||
XM_015151075.2 | XP_015006561.2 | 3191 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |