Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114113.1 | NP_001107585.1 | 310 | 92 | ||
GenBank mRNA | EU340328.1 | - | 310 | 92 |