Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194744.3 | NP_001181673.1 | 91 | 84 |
Gene Symbol: LOC114669749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141587.2 | XP_014997073.1 | 210 | 84 | ||
XM_028842636.1 | XP_028698469.1 | 210 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |