Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161449.1 | NP_001154921.1 | 1551 | 70 | ||
XM_028844729.1 | XP_028700562.1 | 1906 | 70 | |||
XM_028844730.1 | XP_028700563.1 | 1639 | 70 | |||
XM_028844731.1 | XP_028700564.1 | 1228 | 70 | |||
GenBank mRNA | FJ914626.1 | - | 1551 | 70 | ||
JV584082.1 | - | 813 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |