Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166011.3 | NP_001159483.1 | 75 | 54 | ||
XM_028827383.1 | XP_028683216.1 | 732 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |