Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171189.1 | NP_001164660.1 | 409 | 107 | ||
GenBank mRNA | CN646978.1 | - | 221 | 107 | ||
JU320278.1 | - | 495 | 107 | |||
JU320279.1 | - | 495 | 107 | |||
JU472738.1 | - | 485 | 107 | |||
JV043669.1 | - | 477 | 107 | |||
JV528518.1 | - | 483 | 107 | |||
JV566462.1 | - | 467 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |