Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSGALNACT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172170.1 | NP_001165641.1 | 1194 | 96 | ||
XM_028853364.1 | XP_028709197.1 | 1250 | 96 | |||
XM_028853365.1 | XP_028709198.1 | 1250 | 96 | |||
XM_028853366.1 | XP_028709199.1 | 1250 | 96 | |||
XM_028853367.1 | XP_028709200.1 | 1250 | 96 | |||
GenBank mRNA | JU321709.1 | - | 1197 | 96 | ||
JU473204.1 | - | 1187 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |