Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSGALNACT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172170.1 | NP_001165641.1 | 1300 | 104 | ||
XM_028853364.1 | XP_028709197.1 | 1356 | 104 | |||
GenBank mRNA | JU321709.1 | - | 1303 | 104 | ||
JU473204.1 | - | 1293 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |