Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL15RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079517.1 | NP_001072985.1 | 520 | 74 | ||
NM_001172844.1 | NP_001166315.1 | 619 | 74 | |||
XM_015146369.2 | XP_015001855.2 | 618 | 74 | |||
XM_015146370.2 | XP_015001856.1 | 519 | 74 | |||
XM_015146371.2 | XP_015001857.1 | 942 | 74 | |||
XM_028853491.1 | XP_028709324.1 | 843 | 74 | |||
GenBank mRNA | EF033114.1 | - | 619 | 74 | ||
EF033115.1 | - | 520 | 74 | |||
EF033116.1 | - | 421 | 74 | |||
FJ222743.1 | - | 538 | 74 | |||
JU485931.1 | - | 398 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |