Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193929.2 | NP_001180858.2 | 1781 | 87 | ||
XM_028848051.1 | XP_028703884.1 | 2083 | 87 | |||
XM_028848052.1 | XP_028703885.1 | 1805 | 87 | |||
XM_028848053.1 | XP_028703886.1 | 2083 | 87 | |||
XM_028848054.1 | XP_028703887.1 | 2083 | 87 | |||
XM_028848055.1 | XP_028703888.1 | 2083 | 87 | |||
XM_028848056.1 | XP_028703889.1 | 2083 | 87 | |||
XM_028848057.1 | XP_028703890.1 | 2083 | 87 | |||
XM_028848058.1 | XP_028703891.1 | 2083 | 87 | |||
XM_028848059.1 | XP_028703892.1 | 2083 | 87 | |||
XM_028848060.1 | XP_028703893.1 | 1519 | 87 | |||
XM_028848061.1 | XP_028703894.1 | 1209 | 87 |