Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194056.1 | NP_001180985.1 | 1132 | 59 | ||
GenBank mRNA | JV068471.1 | - | 1086 | 59 | ||
JV635684.1 | - | 1146 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |