Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP8-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193418.1 | NP_001180347.1 | 252 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |