Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193441.1 | NP_001180370.1 | 103 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |