Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194133.3 | NP_001181062.1 | 482 | 77 | ||
GenBank mRNA | JU321629.1 | - | 345 | 77 | ||
JV490395.1 | - | 336 | 77 | |||
JV623909.1 | - | 352 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |