Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194250.2 | NP_001181179.1 | 403 | 85 | ||
GenBank mRNA | JU327678.1 | - | 224 | 85 | ||
JU479250.1 | - | 227 | 85 | |||
JV046038.1 | - | 308 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |