Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRGX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194598.1 | NP_001181527.1 | 540 | 63 | ||
XM_028853539.1 | XP_028709372.1 | 625 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |