Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRND | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194688.1 | NP_001181617.1 | 349 | 73 | ||
XM_015149366.2 | XP_015004852.1 | 452 | 73 | |||
GenBank mRNA | JV043458.1 | - | 355 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |